5-Amino-2-nitrobenzoic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Nitro-5-aminobenzoic acid, NSC 74455, 5-Amino-2-nitrobenzoic Acid, 3-Carboxy-4-nitroaniline
L16225-06
L16225-14
CAAAAL16225-03EA
89.92
CAD
CAAAAL16225-03
CAAAAL16225-06
CAAAAL16225-14
5-Amino-2-nitrobenzoic acid 95%
5-Amino-2-nitrobenzoic acid
Purity: 95%
CAS No.: 13280-60-9
Molecular Formula: C7H6N2O4
Formula Weight: 182.14
Melting Point: ca 240° dec.
Beilstein Reference: 2107512
Formula:
C₇H₆N₂O₄ Boiling Pt: 481 °C (1013 hPa) Melting Pt: ∼ 240 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00035748 CAS Number: 13280-60-9 EINECS: 236-283-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...