4-Methyl-2-pyridone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16188-03
L16188-06
L16188-14
CAAAAL16188-03EA
48.66
CAD
CAAAAL16188-03
CAAAAL16188-06
CAAAAL16188-14
4-Methyl-2-pyridone 98+%
4-Methyl-2-pyridone
Purity: 98+%
CAS No.: 13466-41-6
Molecular Formula: C6H7NO
Formula Weight: 109.13
Odor: Odorless
Melting Point: 128-131°
Boiling Point: 186-187°/12mm
Beilstein Reference: 107082
MDL No.: MFCD00134298
Formula:
C₆H₇NO Boiling Pt: 186…187 °C/12 mm Melting Pt: 128…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134298 CAS Number: 13466-41-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...