1-Benzyloxy-4-iodobenzene 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(Benzyloxy)-4-iodobenzene, 4-Benzyloxyiodobenzene

L16092-06 L16092-14
CAAAAL16092-06EA 116.29 CAD
CAAAAL16092-06 CAAAAL16092-14
1-Benzyloxy-4-iodobenzene 98+%
1-Benzyloxy-4-iodobenzene

Purity: 98+% CAS No.: 19578-68-8 Molecular Formula: C13H11IO Formula Weight: 310.14 Melting Point: 60-64° Boiling Point: 140°/1mm MDL No.: MFCD00079704

Formula: C₁₃H₁₁IO
MW: 310.13 g/mol
Boiling Pt: 140 °C/1 mm
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00079704
CAS Number: 19578-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR