Lithium amide 95%

Supplier: Thermo Scientific Chemicals
Danger

L16023-14 L16023-22 L16023-36
CAAAAL16023-14EA 59.41 CAD
CAAAAL16023-14 CAAAAL16023-22 CAAAAL16023-36
Lithium amide 95%
Lithium amide

Purity: 95% CAS No.: 7782-89-0 Molecular Formula: H2LiN Formula Weight: 22.96 Melting Point: 373° Boiling Point: 430° Merck Reference: 14,5522 UN No.: 1390 MDL No.: MFCD00011093

Formula: H₂LiN
Boiling Pt: 430 °C
Melting Pt: 373 °C
Density: 1.178
Storage Temperature: Ambient
MDL Number: MFCD00011093
CAS Number: 7782-89-0
EINECS: 231-968-4
UN: 1390
ADR: 4.3,II
Merck Index: 14,5522

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR