2,6-Dimethyl-4-heptanone ≥90%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16015-0F
L16015-AP
CAAAAL16015-0FEA
125.45
CAD
CAAAAL16015-0F
CAAAAL16015-AP
2,6-Dimethyl-4-heptanone ≥90%
2,6-Dimethyl-4-heptanone
Purity:
Formula:
C₉H₁₈O Boiling Pt: 163…173 °C Melting Pt: –42 °C Density: 0.809 Flash Pt: 47 °C (117 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008940 CAS Number: 108-83-8 EINECS: 203-620-1 UN: 1157 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...