4-Iodophenyl isocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

L16000-03 L16000-06
CAAAAL16000-03EA 73.15 CAD
CAAAAL16000-03 CAAAAL16000-06
4-Iodophenyl isocyanate 97%
4-Iodophenyl isocyanate

Purity: 97% CAS No.: 15845-62-2 Molecular Formula: C7H4INO Formula Weight: 245.02 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Odor: Pungent Melting Point: 43-47° Boiling Point: 84°/5mm Beilstein Reference: 2716151 UN No.: 2811 MDL No.: MFCD00013875

Formula: C₇H₄INO
Boiling Pt: 84 °C/5 mm
Melting Pt: 43…47 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013875
CAS Number: 15845-62-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR