2,5-Dimethylphenyl isocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L15999-03
L15999-06
CAAAAL15999-03EA
37.6
CAD
CAAAAL15999-03
CAAAAL15999-06
2,5-Dimethylphenyl isocyanate 97%
2,5-Dimethylphenyl isocyanate
Purity: 97%
CAS No.: 40397-98-6
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Flash Point: 90°(194°F)
Refractive Index: 1.5315
Boiling Point: 210°
Beilstein Reference: 1100841
UN No.: 2206
MDL No.: MFCD00013854
Formula:
C₉H₉NO Boiling Pt: 210 °C Density: 1.043 Flash Pt: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013854 CAS Number: 40397-98-6 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...