2,5-Difluorophenyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15996-03
L15996-MD
CAAAAL15996-03EA
67.94
CAD
CAAAAL15996-03
CAAAAL15996-MD
2,5-Difluorophenyl isocyanate 98%
2,5-Difluorophenyl isocyanate
Purity: 98%
CAS No.: 39718-32-6
Molecular Formula: C7H3F2NO
Formula Weight: 155.11
Storage Temperature: 2°C to 8°C
Flash Point: 56°(132°F)
Refractive Index: 1.4940
Boiling Point: 78°/33mm
Beilstein Reference: 2091753
UN No.: 3080
MDL No.: MFCD00013843
Formula:
C₇H₃F₂NO Boiling Pt: 78 °C/33 mm Density: 1.323 Flash Pt: 56 °C (132 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013843 CAS Number: 39718-32-6 UN: 3080 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...