3-Methoxy-3-methyl-1-butanol 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methyl-3-methoxybutanol, Solvant 39, MMB, 3-Methoxy-3-methylbutan-1-ol

L15953-0F L15953-AE L15953-AP
CAAAAL15953-0FEA 437.66 CAD
CAAAAL15953-0F CAAAAL15953-AE CAAAAL15953-AP
3-Methoxy-3-methyl-1-butanol 98+%
3-Methoxy-3-methyl-1-butanol

Purity: 98+% CAS No.: 56539-66-3 Molecular Formula: C6H14O2 Formula Weight: 118.18 Flash Point: 71°(159°F) Refractive Index: 1.4280 Boiling Point: 173-175° Beilstein Reference: 1732882 MDL No.: MFCD00044771

Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 173…175 °C
Melting Pt: -50
Density: 0.926
Flash Pt: 71 °C(159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00044771
CAS Number: 56539-66-3
EINECS: 260-252-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR