5-Methylthiazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15879-06
L15879-14
CAAAAL15879-03EA
194.15
CAD
CAAAAL15879-03
CAAAAL15879-06
CAAAAL15879-14
5-Methylthiazole 97%
5-Methylthiazole
Purity: 97%
CAS No.: 3581-89-3
Molecular Formula: C4H5NS
Formula Weight: 99.16
Flash Point: 42°(107°F)
Refractive Index: 1.5280
Odor: Stench
Boiling Point: 141-142°
Beilstein Reference: 104674
UN No.: 1993
MDL No.: MFCD00005335
Formula:
C₄H₅NS Boiling Pt: 141…142 °C Melting Pt: –40.4 °C Density: 1.120 Flash Pt: 42 °C (107 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005335 CAS Number: 3581-89-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...