2-Amino-4-methyl-3-nitropyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Methyl-3-nitro-2-pyridylamine, 4-Methyl-3-nitropyridin-2-amine, 2-Amino-3-nitro-4-picoline
L15841-06
L15841-14
CAAAAL15841-03EA
65.32
CAD
CAAAAL15841-03
CAAAAL15841-06
CAAAAL15841-14
2-Amino-4-methyl-3-nitropyridine 97%
2-Amino-4-methyl-3-nitropyridine
Purity: 97%
CAS No.: 6635-86-5
Molecular Formula: C6H7N3O2
Formula Weight: 153.14
Melting Point: 134-138°
Beilstein Reference: 139111
Formula:
C₆H₇N₃O₂ MW: 153.14 g/mol Melting Pt: 134…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006315 CAS Number: 6635-86-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...