Zinc cyanide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc (II) cyanide

L15836-0B L15836-14 L15836-22
CAAAAL15836-0BEA 237 CAD
CAAAAL15836-0B CAAAAL15836-14 CAAAAL15836-22
Zinc cyanide 98%
Zinc cyanide

Purity: 98% CAS No.: 557-21-1 Molecular Formula: Zn(CN)2 Formula Weight: 117.42 Melting Point: 800° dec. Beilstein Reference: 4124366 Merck Reference: 14,10135 UN No.: 1713 MDL No.: MFCD00011292

Formula: C₂N₂Zn
MW: 117.43
Melting Pt: 800 °C (dec.)
Density: 1.852
Storage Temperature: Ambient
MDL Number: MFCD00011292
CAS Number: 557-21-1
EINECS: 209-162-9
UN: 1713
ADR: 6.1 I
Merck Index: 14,10135

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR