2-Hydroxy-m-toluic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3-Cresotic acid, 2-Hydroxy-3-methylbenzoic acid, 3-Methylsalicylic acid

L15825-14 L15825-22
CAAAAL15825-14EA 33.81 CAD
CAAAAL15825-14 CAAAAL15825-22
2-Hydroxy-m-toluic acid 99%
2-Hydroxy-m-toluic acid

Purity: 99% CAS No.: 83-40-9 Molecular Formula: C8H8O3 Formula Weight: 152.15 Odor: Odorless Melting Point: 163-167° Beilstein Reference: 2086811 Merck Reference: 14,2581 MDL No.: MFCD00002448

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 163…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00002448
CAS Number: 83-40-9
EINECS: 201-473-8
Merck Index: 14,2581

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR