Dipropylamine 99%

Supplier: Thermo Scientific Chemicals
Danger

L15808-AE L15808-AP
CAAAAL15808-AEEA 47.87 CAD
CAAAAL15808-AE CAAAAL15808-AP
Dipropylamine 99%
Dipropylamine

Purity: 99% CAS No.: 142-84-7 Molecular Formula: C6H15N Formula Weight: 101.19 Flash Point: 7°(45°F) Refractive Index: 1.4050 Odor: Ammonia-like Melting Point: -63° Boiling Point: 109-110° Beilstein Reference: 505974 Merck Reference: 14,3343 UN No.: 2383 MDL No.: MFCD00009362

Formula: C₆H₁₅N
Boiling Pt: 109…110 °C
Melting Pt: –63 °C
Density: 0.738
Flash Pt: 7 °C (45 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009362
CAS Number: 142-84-7
EINECS: 205-565-9
UN: 2383
ADR: 3,II
Merck Index: 14,3343

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR