Dimethyl sebacate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,10-Decanedioic acid dimethyl ester, Dimethyl decanedioate, Sebacic acid dimethyl ester

L15805-22 L15805-36
CAAAAL15805-22EA 88.48 CAD
CAAAAL15805-22 CAAAAL15805-36
Dimethyl sebacate 97%
Dimethyl sebacate

Purity: 97% CAS No.: 106-79-6 Molecular Formula: CH3O2C(CH2)8CO2CH3 Formula Weight: 230.31 Flash Point: 145°(293°F) Melting Point: 25-28° Boiling Point: 287-289° Beilstein Reference: 1785523 MDL No.: MFCD00008472

Formula: C₁₂H₂₂O₄
MW: 230.3 g/mol
Boiling Pt: 102
Melting Pt: 38
Density: 0.988
Flash Pt: 145 °C(293 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008472
CAS Number: 106-79-6
EINECS: 203-431-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR