1,6-Dihydroxynaphthalene 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Naphthalene-1,6-diol

L15801-14 L15801-22 L15801-36
CAAAAL15801-14EA 70.15 CAD
CAAAAL15801-14 CAAAAL15801-22 CAAAAL15801-36
1,6-Dihydroxynaphthalene 97+%
1,6-Dihydroxynaphthalene

Purity: 97+% CAS No.: 575-44-0 Molecular Formula: C10H8O2 Formula Weight: 160.17 Melting Point: 136-140° Beilstein Reference: 1939032

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00003981
CAS Number: 575-44-0
EINECS: 209-386-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR