4-Nitro-α,α,α-trifluorotoluene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Nitro-alpha,alpha,alpha-trifluorotoluene, 4-Nitrobenzotrifluoride, 1-Nitro-4-(trifluoromethyl)benzene

L15766-03 L15766-06
CAAAAL15766-03EA 51.35 CAD
CAAAAL15766-03 CAAAAL15766-06
4-Nitro-α,α,α-trifluorotoluene 98%
4-Nitro-α,α,α-trifluorotoluene

Purity: 98% CAS No.: 402-54-0 Molecular Formula: C7H4F3NO2 Formula Weight: 191.11 Flash Point: 88°(190°F) Odor: Characteristic Boiling Point: 81-83°/10mm UN No.: 3431 MDL No.: MFCD00007358

Formula: C₇H₄F₃NO₂
MW: 191.11
Boiling Pt: 204
Melting Pt: 38…42 °C
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007358
CAS Number: 402-54-0
EINECS: 206-948-3
UN: 3431
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR