3-(Hydroxymethyl)indole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Indolmethanol, 3-Indolcarbinol, (1H-Indol-3-yl)methanol, Indole-3-methanol, (Indole-3-yl)methanol, 3-Indolemethanol, Indole-3-carbinol

L15762-03 L15762-06
CAAAAL15762-03EA 42.19 CAD
CAAAAL15762-03 CAAAAL15762-06
3-(Hydroxymethyl)indole 97%
3-(Hydroxymethyl)indole

Purity: 97% CAS No.: 700-06-1 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 96-99° Beilstein Reference: 121323

Formula: C₉H₉NO
MW: 147.18 g/mol
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00005632
CAS Number: 700-06-1
EINECS: 211-836-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR