2,2'-Methyliminodiethanol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15712-22
L15712-36
CAAAAL15712-22EA
50.09
CAD
CAAAAL15712-22
CAAAAL15712-36
2,2'-Methyliminodiethanol 98+%
2,2'-Methyliminodiethanol
Purity: 98+%
CAS No.: 105-59-9
Molecular Formula: C5H13NO2
Formula Weight: 119.17
Flash Point: 127°(260°F)
Refractive Index: 1.4690
Odor: Light
Melting Point: -21°
Boiling Point: 243°
Beilstein Reference: 1734441
MDL No.: MFCD00002848
Formula:
H₃CN(CH₂CH₂OH)₂ Boiling Pt: 243 °C Melting Pt: –21 °C Density: 1.040 Flash Pt: 127 °C(260 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002848 CAS Number: 105-59-9 EINECS: 203-312-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...