Thionyl bromide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L15648-14
L15648-22
CAAAAL15648-14EA
97.8
CAD
CAAAAL15648-14
CAAAAL15648-22
Thionyl bromide 97%
Thionyl bromide
Purity: 97%
CAS No.: 507-16-4
Molecular Formula: Br2OS
Formula Weight: 207.88
Storage Temperature: 2°C to 8°C
Refractive Index: 1.6750
Odor: Acrid
Melting Point: -52°
Boiling Point: 47-49°/20mm
Merck Reference: 14,9347
UN No.: 3264
MDL No.: MFCD00011448
Formula:
Br₂OS Boiling Pt: 47…49 °C/20 mm Melting Pt: –52 °C Density: 2.683 Storage Temperature: Ambient |
MDL Number:
MFCD00011448 CAS Number: 507-16-4 EINECS: 208-064-3 UN: 3264 ADR: 8,III Merck Index: 14,9347 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...