4-Phenylbutyrophenone 98%

Supplier: Thermo Scientific Chemicals

L15597-03 L15597-06 L15597-14
CAAAAL15597-03EA 49.77 CAD
CAAAAL15597-03 CAAAAL15597-06 CAAAAL15597-14
4-Phenylbutyrophenone 98%
4-Phenylbutyrophenone

Purity: 98% CAS No.: 5407-91-0 Molecular Formula: C16H16O Formula Weight: 224.30 Melting Point: 54-56° Boiling Point: 180°/8mm MDL No.: MFCD00022005

Formula: (C₆H₅)C(O)CH₂CH₂CH₂(C₆H₅)
Boiling Pt: 180 °C/8 mm
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00022005
CAS Number: 5407-91-0
EINECS: 226-471-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR