Diisobutylamine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L15495-AP
L15495-AU
CAAAAL15495-APEA
125.77
CAD
CAAAAL15495-AP
CAAAAL15495-AU
Diisobutylamine 99%
Diisobutylamine
Purity: 99%
CAS No.: 110-96-3
Molecular Formula: C8H19N
Formula Weight: 129.25
Flash Point: 30°(86°F)
Refractive Index: 1.4090
Melting Point: -77°
Boiling Point: 137-139°
Beilstein Reference: 1209251
UN No.: 2361
MDL No.: MFCD00008930
Formula:
C₈H₁₉N Boiling Pt: 137…139 °C Melting Pt: –77 °C Density: 0.744 Flash Pt: 30 °C (86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008930 CAS Number: 110-96-3 EINECS: 203-819-3 UN: 2361 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...