3',5'-Dimethylacetophenone 97%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
1-(3,5-Dimethylphenyl)ethanone, 1-(3,5-Dimethylphenyl)ethan-1-one
L15480-03
L15480-06
CAAAAL15480-03EA
92.59
CAD
CAAAAL15480-03
CAAAAL15480-06
3',5'-Dimethylacetophenone 97%
3',5'-Dimethylacetophenone
Purity: 97%
CAS No.: 5379-16-8
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 93°(199°F)
Refractive Index: 1.5280
Odor: Characteristic
Melting Point: ca 20°
Boiling Point: 238°
Beilstein Reference: 2240987
MDL No.: MFCD01075693
Formula:
C₁₀H₁₂O MW: 148.2 g/mol Boiling Pt: 238 °C Melting Pt: ∼ 20 °C Density: 0.988 Flash Pt: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01075693 CAS Number: 5379-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...