2-Methoxy-1-methylethyl acetate (PGMEA) 99% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Propylene glycol 1-methyl ether 2-acetat, 2-(1-Methoxy)propyl acetate, 1-Methoxy-2-propyl acetate, Propylene glycol methyl ether acetat, 1,2-Propanediol monomethyl ether acetate, DOWANOL PMA
Stabilization:
50 ppm Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Purity: 99%
CAS No.: 108-65-6
Molecular Formula: C6H12O3
Formula Weight: 132.16
Flash Point: 43°(109°F)
Refractive Index: 1.4020
Odor: Ester-like
pH: 4
Melting Point: -67°
Boiling Point: 145-146°
Beilstein Reference: 1751656
UN No.: 3272
MDL No.: MFCD00038500
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol Boiling Pt: 145.8 Melting Pt: -67 Density: 0.968 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00038500 CAS Number: 108-65-6 EINECS: 203-603-9 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...