Octadecylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L15458-14
L15458-22
CAAAAL15458-14EA
173.8
CAD
CAAAAL15458-14
CAAAAL15458-22
Octadecylamine 97%
Octadecylamine
Purity: 97%
CAS No.: 124-30-1
Molecular Formula: C18H39N
Formula Weight: 269.52
Flash Point: 148°(298°F)
Melting Point: 53-55°
Boiling Point: 180-183°/5mm
Beilstein Reference: 636111
MDL No.: MFCD00008159
Formula:
H₃C(CH₂)₁₆CH₂NH₂ Boiling Pt: 180…183 °C/5 mm Melting Pt: 53…55 °C Density: 0.862 Flash Pt: 148 °C (298 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008159 CAS Number: 124-30-1 EINECS: 204-695-3 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...