(±)-1-Aminopropan-2-ol 94% remainder 2-amino-1-propanol
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aminopropanol, Hydroxypropylamine, 1-Aminopropan-2-ol, DL-1-Amino-2-propanol, (±)-Isopropanolamine, Aminoisopropyl alcohol, iso-propanol amine Amino-2-propanol DL-Isopropanolamine (±)-1-Amino-2-propanol (±)-Isopropanolamine
L15343-0B
L15343-0E
L15343-30
CAAAAL15343-0BEA
72.36
CAD
CAAAAL15343-0B
CAAAAL15343-0E
CAAAAL15343-30
(±)-1-Aminopropan-2-ol 94% remainder 2-amino-1-propanol
(±)-1-Aminopropan-2-ol
Purity: 94%
CAS No.: 78-96-6
Molecular Formula: C3H9NO
Formula Weight: 75.11
Flash Point: 71°(159°F)
Refractive Index: 1.4480
Boiling Point: 160°
Beilstein Reference: 605275
UN No.: 2735
MDL No.: MFCD00008139
Formula:
C₃H₉NO Boiling Pt: 160 °C Melting Pt: 1 °C Density: 0.959 Flash Pt: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008139 CAS Number: 78-96-6 EINECS: 201-162-7 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...