3-(2-Chlorophenyl)propionic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(2-Chlorophenyl)propanoic acid

L15306-03 L15306-06
CAAAAL15306-03EA 94.33 CAD
CAAAAL15306-03 CAAAAL15306-06
3-(2-Chlorophenyl)propionic acid 98+%
3-(2-Chlorophenyl)propionic acid

Purity: 98+% CAS No.: 1643-28-3 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Melting Point: 94-97° Boiling Point: 171°/10mm MDL No.: MFCD00016547

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Boiling Pt: 171 °C/10 mm
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00016547
CAS Number: 1643-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR