m-Tolyl isocyanate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15255-06
L15255-14
CAAAAL15255-06EA
54.04
CAD
CAAAAL15255-06
CAAAAL15255-14
m-Tolyl isocyanate 99%
m-Tolyl isocyanate
Purity: 99%
CAS No.: 621-29-4
Molecular Formula: C8H7NO
Formula Weight: 133.15
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Refractive Index: 1.5305
Odor: Pungent
Boiling Point: 75-76°/12mm
Beilstein Reference: 386149
UN No.: 2206
MDL No.: MFCD00002021
Formula:
C₈H₇NO Boiling Pt: 75…76 °C/12 mm Melting Pt: 195…198 °C Density: 1.033 Flash Pt: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002021 CAS Number: 621-29-4 EINECS: 210-676-0 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...