2-(4-Aminophenyl)ethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

L15223-06 L15223-14 L15223-22
CAAAAL15223-06EA 67.78 CAD
CAAAAL15223-06 CAAAAL15223-14 CAAAAL15223-22
2-(4-Aminophenyl)ethanol 97%
2-(4-Aminophenyl)ethanol

Purity: 97% CAS No.: 104-10-9 Molecular Formula: C8H11NO Formula Weight: 137.18 Melting Point: 108-110° Beilstein Reference: 907205

Formula: C₈H₁₁NO
Boiling Pt: 235 - 255 °C (1013 hPa)
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007922
CAS Number: 104-10-9
EINECS: 203-174-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR