L(-)-Fenchone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(1R)-(-)-Fenchone
L15151-18
L15151-30
CAAAAL15151-18EA
64.46
CAD
CAAAAL15151-18
CAAAAL15151-30
L(-)-Fenchone 98+%
L(-)-Fenchone
Purity: 98+%
CAS No.: 7787-20-4
Molecular Formula: C10H16O
Formula Weight: 152.24
Flash Point: 73°(163°F)
Refractive Index: 1.4630
Melting Point: 3-5°
Boiling Point: 192-194°
Beilstein Reference: 2042710
Optical Rotation: -52° (neat)
MDL No.: MFCD00151104
Formula:
C₁₀H₁₆O Boiling Pt: 192…194 °C Melting Pt: 3…5 °C Density: 0.945 Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00151104 CAS Number: 7787-20-4 EINECS: 232-107-5 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...