Pyridinium dichromate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: PDC

L15132-14 L15132-22
CAAAAL15132-14EA 57.35 CAD
CAAAAL15132-14 CAAAAL15132-22 CAAAAL15132-36
Pyridinium dichromate 98%
Pyridinium dichromate

Purity: 98% CAS No.: 20039-37-6 Molecular Formula: C10H12Cr2N2O7 Formula Weight: 376.21 Melting Point: ca 145-147° UN No.: 3087 MDL No.: MFCD00013105

Formula: C₁₀N₁₂Cr₂N₂O₇
MW: 532.19 g/mol
Melting Pt: ∼ 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00013105
CAS Number: 20039-37-6
EINECS: 243-478-8
UN: 3085
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR