2-Iodobenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals
Warning

L15126-06 L15126-14
CAAAAL15126-06EA 63.99 CAD
CAAAAL15126-06 CAAAAL15126-14
2-Iodobenzyl alcohol 99%
2-Iodobenzyl alcohol

Purity: 99% CAS No.: 5159-41-1 Molecular Formula: C7H7IO Formula Weight: 234.04 Melting Point: 89-92° Beilstein Reference: 2079487

Formula: C₇H₇IO
MW: 234.04 g/mol
Boiling Pt: 145
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00004610
CAS Number: 5159-41-1
EINECS: 225-933-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR