(4-Iodophenyl)acetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-Iodophenyl)acetonitrile, 4-Iodobenzyl cyanide, 4-Iodophenylacetonitrile

L15119-03 L15119-06
CAAAAL15119-03EA 104.28 CAD
CAAAAL15119-03 CAAAAL15119-06
(4-Iodophenyl)acetonitrile 97%
(4-Iodophenyl)acetonitrile

Purity: 97% CAS No.: 51628-12-7 Molecular Formula: C8H6IN Formula Weight: 243.05 Flash Point: 110°(230°F) Melting Point: 52-54° Beilstein Reference: 1934479 UN No.: 3439 MDL No.: MFCD00060306

Formula: C₈H₆IN
MW: 243.05 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 52…54 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060306
CAS Number: 51628-12-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR