(3-Iodophenyl)acetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

L15118-03 L15118-06
CAAAAL15118-03EA 103.96 CAD
CAAAAL15118-03 CAAAAL15118-06
(3-Iodophenyl)acetonitrile 97%
(3-Iodophenyl)acetonitrile

Purity: 97% CAS No.: 130723-54-5 Molecular Formula: C8H6IN Formula Weight: 243.05 Flash Point: 110°(230°F) Melting Point: 33-35° Boiling Point: 138°/12mm Beilstein Reference: 7350695 UN No.: 3439 MDL No.: MFCD00040890

Formula: C₈H₆IN
Boiling Pt: 138 °C/12 mm
Melting Pt: 33…35 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040890
CAS Number: 130723-54-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR