1-Hexadecanethiol, (max. 4% H₂O) 97% (dry weight)
Supplier: Thermo Scientific Chemicals
L15099-AC
L15099-AE
L15099-AP
CAAAAL15099-ACEA
68.89
CAD
CAAAAL15099-AC
CAAAAL15099-AE
CAAAAL15099-AP
1-Hexadecanethiol, (max. 4% H₂O) 97% (dry weight)
1-Hexadecanethiol
Purity: 97%
CAS No.: 2917-26-2
Molecular Formula: C16H34S
Formula Weight: 258.51
Flash Point: 135°(275°F)
Refractive Index: 1.4625
Odor: Unpleasant
Melting Point: 19-23°
Boiling Point: 334°
Beilstein Reference: 1748495
MDL No.: MFCD00011677
Formula:
C₁₆H₃₄S Boiling Pt: 334 °C Melting Pt: 19…23 °C Density: 0.846 Flash Pt: 135 °C(275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011677 CAS Number: 2917-26-2 EINECS: 220-846-6 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...