3-Pyridineboronic acid
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15040-03
L15040-06
CAAAAL15040-03EA
113.44
CAD
CAAAAL15040-03
CAAAAL15040-06
3-Pyridineboronic acid
3-Pyridineboronic acid
CAS No.: 1692-25-7
Molecular Formula: C5H6BNO2
Formula Weight: 122.92
Storage Temperature: 2°C to 8°C
Melting Point:
Formula:
C₅H₆BNO₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00674177 CAS Number: 1692-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...