Polysorbate 60
Supplier: Thermo Scientific Chemicals
Synonyms:
Sorbitan monostearate, ethoxylated
, Polyoxyethylene sorbitan monostearate
L15031-AE
L15031-AP
L15031-AP
L15031-AE
CAAAAL15031-AEEA
39.5
CAD
CAAAAL15031-AE
AAAL15031-AE
AAAL15031-AP
CAAAAL15031-AP
Polysorbate 60
Polysorbate 60
Formula:
C₆₄H₁₂₆O₂₆ Density: 1.080 Flash Pt: >150 °C (302 °F) |
MDL Number:
MFCD01780110 CAS Number: 9005-67-8 Merck Index: 14,7582 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...