Polysorbate 40

Supplier: Thermo Scientific Chemicals

Synonyms: Sorbitan monopalmitate, ethoxylated, Polyoxyethylene sorbitan monopalmitate

L15030-AE L15030-AP
CAAAAL15030-AEEA 35.55 CAD
CAAAAL15030-AE CAAAAL15030-AP
Polysorbate 40
Polysorbate 40

CAS No.: 9005-66-7 Molecular Formula: C62H122O26 Refractive Index: 1.4700 MDL No.: MFCD00165345

Formula: C₆₂H₁₂₂O₂₆
Density: 1.090
Storage Temperature: Ambient
MDL Number: MFCD00165345
CAS Number: 9005-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR