Polysorbate 20

Supplier: Thermo Scientific Chemicals

Synonyms: Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated

L15029-AE L15029-AP
CAAAAL15029-AEEA 44.87 CAD
CAAAAL15029-AE CAAAAL15029-AP
Polysorbate 20
Polysorbate 20

Used for solubilizing membrane proteins during isolation and purification


CAS No.: 9005-64-5 Molecular Formula: C58H114O26 Flash Point: 110°(230°F) Refractive Index: 1.4700 Boiling Point: 100° MDL No.: MFCD00165986

Formula: C₅₈H₁₁₄O₂₆
Boiling Pt: >100 °C
Density: 1.105
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR