Cyclooctane 99+%

Supplier: Thermo Scientific Chemicals
Danger

L15000-AD L15000-AK
CAAAAL15000-ADEA 63.52 CAD
CAAAAL15000-AD CAAAAL15000-AK
Cyclooctane 99+%
Cyclooctane

Purity: 99+% CAS No.: 292-64-8 Molecular Formula: C8H16 Formula Weight: 112.22 Flash Point: 28°(82°F) Refractive Index: 1.4580 Melting Point: 12-14° Boiling Point: 150-152° Beilstein Reference: 1900349 UN No.: 3295 MDL No.: MFCD00004162

Formula: C₈H₁₆
Boiling Pt: 150…152 °C
Melting Pt: 12…14 °C
Density: 0.835
Flash Pt: 28 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004162
CAS Number: 292-64-8
EINECS: 206-031-8
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR