Glycolic acid ∼66 - 70% in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14980-0F
L14980-AP
CAAAAL14980-0FEA
116.92
CAD
CAAAAL14980-0F
CAAAAL14980-AP
Glycolic acid ∼66 - 70% in aqueous solution
Glycolic acid
CAS No.: 79-14-1
Molecular Formula: C2H4O3
Formula Weight: 76.05
Flash Point: None
Refractive Index: 1.411
Odor: Sweet
Boiling Point: 112°
Beilstein Reference: 1209322
Merck Reference: 14,4498
UN No.: 3265
MDL No.: MFCD00004312
Formula:
C₂H₄O₃ Boiling Pt: 112 °C Density: 1.270 Storage Temperature: Ambient |
MDL Number:
MFCD00004312 CAS Number: 79-14-1 EINECS: 201-180-5 UN: 3265 ADR: 8,III Merck Index: 14,4498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...