2-Chloro-3-nitrobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14954-03
L14954-06
CAAAAL14954-03EA
71.57
CAD
CAAAAL14954-03
CAAAAL14954-06
2-Chloro-3-nitrobenzoic acid 98%
2-Chloro-3-nitrobenzoic acid
Purity: 98%
CAS No.: 3970-35-2
Molecular Formula: C7H4ClNO4
Formula Weight: 201.57
Odor: Odorless
Melting Point: 182-186°
Beilstein Reference: 645427
MDL No.: MFCD00007069
Formula:
C₇H₄ClNO₄ Melting Pt: 182…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007069 CAS Number: 3970-35-2 EINECS: 223-590-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...