2,6-Difluorophenyl isocyanate 97%

Supplier: Thermo Scientific Chemicals
Warning

L14779-03 L14779-06
CAAAAL14779-03EA 86.11 CAD
CAAAAL14779-03 CAAAAL14779-06
2,6-Difluorophenyl isocyanate 97%
2,6-Difluorophenyl isocyanate

Purity: 97% CAS No.: 65295-69-4 Molecular Formula: C7H3F2NO Formula Weight: 155.11 Flash Point: 58°(136°F) Refractive Index: 1.4928 Melting Point: 22-25° Boiling Point: 60°/10mm Beilstein Reference: 7115873 UN No.: 2930 MDL No.: MFCD00001998

Formula: C₇H₃F₂NO
MW: 155.1 g/mol
Boiling Pt: 60 °C/10 mm
Melting Pt: 22…25 °C
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001998
CAS Number: 65295-69-4
UN: 2478
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR