Chloroform 99+% stabilised ethanol-free
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
Stabilization:
∼50 ppm Stabilised with amylene
L14759-0F
L14759-AP
L14759-AU
CAAAAL14759-0FEA
194.34
CAD
CAAAAL14759-0F
CAAAAL14759-AP
CAAAAL14759-AU
Chloroform 99+% stabilised ethanol-free
Chloroform
Purity: 99+%
CAS No.: 67-66-3
Molecular Formula: CHCl3
Formula Weight: 119.38
Refractive Index: 1.4460
Odor: Characteristic
Boiling Point: 60.5-61.5°
Beilstein Reference: 1731042
Merck Reference: 14,2141
UN No.: 1888
MDL No.: MFCD00000826
Formula:
CHCl₃ MW: 119.38 g/mol Boiling Pt: 61.7 Melting Pt: -63 Density: 1.492 Storage Temperature: Ambient |
MDL Number:
MFCD00000826 CAS Number: 67-66-3 EINECS: 200-663-8 UN: 1888 ADR: 6.1 III REACH: 05-2114614906-45 Merck Index: 14,2141 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...