3,5-Dimethylbenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (3,5-Dimethylphenyl)methanol, 5-(Hydroxymethyl)-m-xylene, 3,5-Dimethylbenzylalcohol

L14726-03 L14726-06 L14726-14
CAAAAL14726-03EA 58.93 CAD
CAAAAL14726-03 CAAAAL14726-06 CAAAAL14726-14
3,5-Dimethylbenzyl alcohol 99%
3,5-Dimethylbenzyl alcohol

Purity: 99% CAS No.: 27129-87-9 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 106°(222°F) Refractive Index: 1.5310 Boiling Point: 218-221° Beilstein Reference: 2077285 MDL No.: MFCD00004648

Formula: C₉H₁₂O
Boiling Pt: 218…221 °C
Flash Pt: 106 °C(222 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004648
CAS Number: 27129-87-9
EINECS: 248-241-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR