Diethoxy(dimethyl)silane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dimethyldiethoxysilane
L14725-0B
L14725-18
L14725-30
CAAAAL14725-0BEA
363.4
CAD
CAAAAL14725-0B
CAAAAL14725-18
CAAAAL14725-30
Diethoxy(dimethyl)silane 97%
Diethoxy(dimethyl)silane
Purity: 97%
CAS No.: 78-62-6
Molecular Formula: C6H16O2Si
Formula Weight: 148.28
Flash Point: 11°(52°F)
Refractive Index: 1.3820
Odor: Mild
Melting Point: -70°
Boiling Point: 111-113°
Beilstein Reference: 1736110
UN No.: 2380
MDL No.: MFCD00009068
Formula:
C₆H₁₆O₂Si MW: 148.28 g/mol Boiling Pt: 114 Melting Pt: -87 Density: 0.865 Flash Pt: 11 °C (52 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009068 CAS Number: 78-62-6 EINECS: 201-127-6 UN: 2380 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...