2-Chloro-6-fluorobenzaldehyde oxime 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14719-03
L14719-06
CAAAAL14719-03EA
28.28
CAD
CAAAAL14719-03
CAAAAL14719-06
2-Chloro-6-fluorobenzaldehyde oxime 97%
2-Chloro-6-fluorobenzaldehyde oxime
Purity: 97%
CAS No.: 443-33-4
Molecular Formula: C7H5ClFNO
Formula Weight: 173.58
Melting Point: 125-127°
Beilstein Reference: 3247886
Formula:
C₇H₅ClFNO Melting Pt: 125…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00837704 CAS Number: 443-33-4 EINECS: 207-135-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...