Polypropylene glycol, M.W. 2,000

Supplier: Thermo Scientific Chemicals

Synonyms: Poly(propylene oxide), PPG

L14699-AE L14699-AP
CAAAAL14699-AEEA 69.99 CAD
CAAAAL14699-AE CAAAAL14699-AP
Polypropylene glycol, M.W. 2,000
Polypropylene glycol

CAS No.: 25322-69-4 Molecular Formula: H[OCH(CH3)CH2]nOH Flash Point: 194°(381°F) Refractive Index: 1.4510 MDL No.: MFCD01779687

Formula: H(OCH₂CH₂CH₂)nOH
Density: 1.002
Flash Pt: 194 °C(381 °F) (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00084446
CAS Number: 25322-69-4
EINECS: 500-039-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR