Petroleum spirit, 40…60 °C

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Petroleum ether, Petroleum benzine

L14649-0F L14649-AU
CAAAAL14649-0FEA 104.28 CAD
CAAAAL14649-0F CAAAAL14649-AU
Petroleum spirit, 40…60 °C
Petroleum spirit (8032-32-4)

CAS No.: 8032-32-4 Flash Point: -40°(-40°F) Odor: Petroleum-like Boiling Point: 40-60° UN No.: 1268 MDL No.: MFCD00081849

Boiling Pt: 40…60 °C
Density: 0.660
Flash Pt: –40 °C (–40 °F)
Storage Temperature: Ambient
CAS Number: 8032-32-4
EINECS: 232-453-7
UN: 1268
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR