4-Quinolinecarboxylic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Quinoline-4-carboxylic acid, NSC 13138

L14611-03 L14611-06
CAAAAL14611-03EA 101.44 CAD
CAAAAL14611-03 CAAAAL14611-06 CAAAAL14611-MD
4-Quinolinecarboxylic acid 98+%
4-Quinolinecarboxylic acid

Purity: 98+% CAS No.: 486-74-8 Molecular Formula: C10H7NO2 Formula Weight: 173.17 Melting Point: 253-255° Beilstein Reference: 5224

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Melting Pt: 253…255 °C
Storage Temperature: Ambient
MDL Number: MFCD00006782
CAS Number: 486-74-8
EINECS: 207-640-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR